Product Details

SNP ID
rs150955614
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:28340341 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCATAGACCGTTGTGGAAAGCAC[A/T]TTGGCACCATTTTGAATTACCTCCG
Phenotype
MIM: 614394 MIM: 611519 MIM: 191161
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
IFT20 PubMed Links

Gene Details

Gene
IFT20
Gene Name
intraflagellar transport 20
There are no transcripts associated with this gene.

Gene
POLDIP2
Gene Name
DNA polymerase delta interacting protein 2
There are no transcripts associated with this gene.

Gene
TNFAIP1
Gene Name
TNF alpha induced protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021137.4 643 Missense Mutation ATT,TTT I80F NP_066960.1
XM_017024993.1 643 Missense Mutation ATT,TTT I80F XP_016880482.1

View Full Product Details