Product Details
- SNP ID
-
rs150985019
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:58193110 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGGCCAAGTTGCTGGTGGATGCTG[C/G]CTACAATTGGACCCAGAAGAGGTGG
- Phenotype
-
MIM: 131399
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
EPX
PubMed Links
Gene Details
- Gene
- EPX
- Gene Name
- eosinophil peroxidase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000502.5 |
272 |
Missense Mutation |
GCC,GGC |
A50G |
NP_000493.1 |
View Full Product Details