Product Details

SNP ID
rs151038855
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:8369387 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGGCTTTATAGCCAAACATTGCC[G/T]CAAATGGACTTTGCTGCAAGGAAAC
Phenotype
MIM: 603704
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KRBA2 PubMed Links

Gene Details

Gene
KRBA2
Gene Name
KRAB-A domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304947.1 2478 Missense Mutation GAG,GCG E327A NP_001291876.1
NM_213597.3 2478 Missense Mutation GAG,GCG E409A NP_998762.1
XM_005256443.2 2478 Missense Mutation GAG,GCG E375A XP_005256500.1
XM_011523643.1 2478 Missense Mutation GAG,GCG E375A XP_011521945.1
XM_011523644.1 2478 Missense Mutation GAG,GCG E375A XP_011521946.1
XM_017024163.1 2478 Missense Mutation GAG,GCG E375A XP_016879652.1
XM_017024164.1 2478 Missense Mutation GAG,GCG E372A XP_016879653.1
XM_017024165.1 2478 Missense Mutation GAG,GCG E327A XP_016879654.1
Gene
LOC100128288
Gene Name
uncharacterized LOC100128288
There are no transcripts associated with this gene.

Gene
RPL26
Gene Name
ribosomal protein L26
There are no transcripts associated with this gene.

View Full Product Details