Product Details

SNP ID
rs138338692
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:59269076 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTCAGACGCAGCGCCGCGATGCT[A/G]CTGTTGCGCGGGTCCGCCTCGTCCA
Phenotype
MIM: 601881
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAX PubMed Links

Gene Details

Gene
RAX
Gene Name
retina and anterior neural fold homeobox
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013435.2 1156 Silent Mutation AGC,AGT S323S NP_038463.2

View Full Product Details