Product Details

SNP ID
rs139330044
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:46105256 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTATTCCACAGTATGAGGTGAACCC[A/C]CGGACCACAGAGATTTTACATCACC
Phenotype
MIM: 164360 MIM: 608775
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ATP5A1 PubMed Links

Gene Details

Gene
ATP5A1
Gene Name
ATP synthase, H+ transporting, mitochondrial F1 complex, alpha subunit 1, cardiac muscle
There are no transcripts associated with this gene.

Gene
HAUS1
Gene Name
HAUS augmin like complex subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138443.3 Intron NP_612452.1

View Full Product Details