Product Details

SNP ID
rs144636863
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:9485411 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAGTTTCATTTCATTGTGATCAG[A/G]GAACATACTTTGTATGATTTTAGTC
Phenotype
MIM: 605801
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RALBP1 PubMed Links

Gene Details

Gene
RALBP1
Gene Name
ralA binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006788.3 Intron NP_006779.1

View Full Product Details