Product Details

SNP ID
rs146667523
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:2544197 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTCTATTTTATAAAAACATACCA[A/G]CAAGCGGTGGCTTATGTGAGTGAAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
METTL4 PubMed Links

Gene Details

Gene
METTL4
Gene Name
methyltransferase like 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308401.1 2064 Intron NP_001295330.1
NM_022840.4 2064 Missense Mutation GCT,GTT A424V NP_073751.3
XM_005258132.3 2064 Intron XP_005258189.1
XM_005258133.2 2064 Missense Mutation GCT,GTT A277V XP_005258190.1
XM_011525730.2 2064 Intron XP_011524032.1

View Full Product Details