Product Details

SNP ID
rs147796465
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:21749583 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTGTCTTTTAATAGAGGAATTTA[A/G]CTTGTTCATATGTATTGTGACAACT
Phenotype
MIM: 608677
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIB1 PubMed Links

Gene Details

Gene
MIB1
Gene Name
mindbomb E3 ubiquitin protein ligase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020774.3 Intron NP_065825.1
XM_011526098.1 Intron XP_011524400.1
XM_017025873.1 Intron XP_016881362.1
XM_017025874.1 Intron XP_016881363.1
XM_017025875.1 Intron XP_016881364.1

View Full Product Details