Product Details

SNP ID
rs150234249
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:12449881 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCATCTGATTTGTCCATAGACTT[A/C]GATTTTGAGGAGAACCTGGGGTGAA
Phenotype
MIM: 609216
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC105371998 PubMed Links

Gene Details

Gene
LOC105371998
Gene Name
uncharacterized LOC105371998
There are no transcripts associated with this gene.

Gene
SPIRE1
Gene Name
spire type actin nucleation factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128626.1 2438 Silent Mutation TCG,TCT S676S NP_001122098.1
NM_001128627.1 2438 Silent Mutation TCG,TCT S542S NP_001122099.1
NM_020148.2 2438 Silent Mutation TCG,TCT S662S NP_064533.3
XM_005258122.4 2438 Silent Mutation TCG,TCT S720S XP_005258179.1
XM_011525701.1 2438 Silent Mutation TCG,TCT S734S XP_011524003.1
XM_011525702.1 2438 Silent Mutation TCG,TCT S614S XP_011524004.1
XM_011525703.2 2438 Intron XP_011524005.1

View Full Product Details