Product Details

SNP ID
rs151128517
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:59329271 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGTAAGAATAATCAGAATGATTC[A/C]CCTTGAATTTCAGCCTGAAAAATGT
Phenotype
MIM: 601567
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LMAN1 PubMed Links

Gene Details

Gene
LMAN1
Gene Name
lectin, mannose binding 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005570.3 3387 UTR 3 NP_005561.1

View Full Product Details