Product Details

SNP ID
rs138553345
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:3771582 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGCCAGCTCCTCACGGCTGTAC[A/C]CATCCGGGTAGTGAGAGGCCTCGAA
Phenotype
MIM: 604262 MIM: 600038 MIM: 611858 MIM: 610362
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
APBA3 PubMed Links

Gene Details

Gene
APBA3
Gene Name
amyloid beta precursor protein binding family A member 3
There are no transcripts associated with this gene.

Gene
MATK
Gene Name
megakaryocyte-associated tyrosine kinase
There are no transcripts associated with this gene.

Gene
MRPL54
Gene Name
mitochondrial ribosomal protein L54
There are no transcripts associated with this gene.

Gene
RAX2
Gene Name
retina and anterior neural fold homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319074.1 229 Missense Mutation GGG,GTG G100V NP_001306003.1
NM_032753.3 229 Missense Mutation GGG,GTG G54V NP_116142.1

View Full Product Details