Product Details

SNP ID
rs138884282
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:56775313 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGTGTTTGTTCTCTTGGGATGCT[A/G]ACTGGGGACTCGTACATATTCCAGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZIM2 PubMed Links

Gene Details

Gene
ZIM2
Gene Name
zinc finger imprinted 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146326.1 1502 Missense Mutation TCA,TTA S320L NP_001139798.1
NM_001146327.1 1502 Missense Mutation TCA,TTA S320L NP_001139799.1
NM_015363.4 1502 Missense Mutation TCA,TTA S320L NP_056178.3
Gene
ZIM2-AS1
Gene Name
ZIM2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details