Product Details

SNP ID
rs139032859
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:37187082 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATACATAGAGTTTTTCTCCTGTA[G/T]GAACTTTCAGATGTACCTTGAACTG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF585B PubMed Links

Gene Details

Gene
ZNF585B
Gene Name
zinc finger protein 585B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152279.3 707 Missense Mutation CAT,CCT H152P NP_689492.3

View Full Product Details