Product Details

SNP ID
rs139076851
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:547305 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCTCCTTTGGGACCCAGATCAT[C/T]GGGGGCCGGGAGGTGATCCCCCACT
Phenotype
MIM: 116948 MIM: 600311
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDC34 PubMed Links

Gene Details

Gene
CDC34
Gene Name
cell division cycle 34
There are no transcripts associated with this gene.

Gene
GZMM
Gene Name
granzyme M
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258351.1 119 UTR 5 NP_001245280.1
NM_005317.3 119 Silent Mutation ATC,ATT I27I NP_005308.1

View Full Product Details