Product Details

SNP ID
rs139281028
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57941396 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGAGGTAGGATTTACAGGTAAAG[A/G]ATTTTCCACATTCACTGCACTCATA
Phenotype
MIM: 606956
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF256 PubMed Links

Gene Details

Gene
ZNF256
Gene Name
zinc finger protein 256
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005773.2 1647 Missense Mutation TCC,TTC S471F NP_005764.2
XM_017026138.1 1647 Missense Mutation TCC,TTC S318F XP_016881627.1
Gene
ZNF418
Gene Name
zinc finger protein 418
There are no transcripts associated with this gene.

View Full Product Details