Product Details
- SNP ID
-
rs139482540
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:9126291 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAATTTTCATGACAGAGGAGACAAT[C/T]CGTGTGTAAGAGAAAATGATCCCAG
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
OR7G3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs61751875] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR7G3
- Gene Name
- olfactory receptor family 7 subfamily G member 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001001958.1 |
660 |
Silent Mutation |
CGA,CGG |
R220R |
NP_001001958.1 |
View Full Product Details