Product Details
- SNP ID
-
rs139591787
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:38304952 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGAAGGAGAAGGGCAAGAAGAAGGA[C/G]GCTCCCCACTGAAGGGCCCTGGACA
- Phenotype
-
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
C19orf33
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs139805446,rs147127327] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C19orf33
- Gene Name
- chromosome 19 open reading frame 33
- Gene
- YIF1B
- Gene Name
- Yip1 interacting factor homolog B, membrane trafficking protein
View Full Product Details