Product Details

SNP ID
rs139644602
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49880269 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCCCCTTACTGTCTGCTCCTTTC[C/T]TCTCCTAAGGACTCCAGTGGGGGTG
Phenotype
MIM: 610221 MIM: 609742 MIM: 616659
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AKT1S1 PubMed Links

Gene Details

Gene
AKT1S1
Gene Name
AKT1 substrate 1
There are no transcripts associated with this gene.

Gene
IL4I1
Gene Name
interleukin 4 induced 1
There are no transcripts associated with this gene.

Gene
MIR4750
Gene Name
microRNA 4750
There are no transcripts associated with this gene.

Gene
TBC1D17
Gene Name
TBC1 domain family member 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168222.1 Intron NP_001161694.1
NM_024682.2 Intron NP_078958.2
XM_011527317.2 Intron XP_011525619.1

View Full Product Details