Product Details

SNP ID
rs139646557
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:4538149 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGGCTGCCCTAGGGATGCCCAGA[A/G]CCCCTCGGGCACGCTGGCCAGTGAG
Phenotype
MIM: 611289 MIM: 613248 MIM: 608873
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRG1 PubMed Links

Gene Details

Gene
LRG1
Gene Name
leucine rich alpha-2-glycoprotein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052972.2 846 Missense Mutation CTC,TTC L279F NP_443204.1
Gene
PLIN5
Gene Name
perilipin 5
There are no transcripts associated with this gene.

Gene
SEMA6B
Gene Name
semaphorin 6B
There are no transcripts associated with this gene.

View Full Product Details