Product Details

SNP ID
rs140137479
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:53237232 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTTATGCTGAGTAAGATTTGAAC[A/G]TTCAGTAAAGGCTTTGCCACATTCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF677 PubMed Links

Gene Details

Gene
ZNF677
Gene Name
zinc finger protein 677
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317998.1 1907 Missense Mutation CGT,TGT R499C NP_001304927.1
NM_182609.3 1907 Missense Mutation CGT,TGT R499C NP_872415.1
XM_005258862.4 1907 Missense Mutation CGT,TGT R463C XP_005258919.1
XM_011526912.2 1907 Missense Mutation CGT,TGT R499C XP_011525214.1

View Full Product Details