Product Details

SNP ID
rs140212178
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48048371 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCAAGCAGCAACTTCGGGCACTA[A/T]CCTTCGGGTAGTAGAGCCTGGGGAG
Phenotype
MIM: 607315 MIM: 603602
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CABP5 PubMed Links

Gene Details

Gene
CABP5
Gene Name
calcium binding protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019855.4 1996 Intron NP_062829.1
XM_017026992.1 1996 Intron XP_016882481.1
Gene
PLA2G4C
Gene Name
phospholipase A2 group IVC
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159322.1 1996 Missense Mutation GAT,GTT D543V NP_001152794.1
NM_001159323.1 1996 UTR 3 NP_001152795.1
NM_003706.2 1996 Missense Mutation GAT,GTT D533V NP_003697.2
XM_011527431.2 1996 Missense Mutation GAT,GTT D515V XP_011525733.1
XM_011527432.2 1996 Intron XP_011525734.1
XM_011527433.2 1996 Intron XP_011525735.1
XM_017027411.1 1996 UTR 3 XP_016882900.1
XM_017027412.1 1996 Missense Mutation GAT,GTT D354V XP_016882901.1
XM_017027413.1 1996 UTR 3 XP_016882902.1
XM_017027414.1 1996 Intron XP_016882903.1

View Full Product Details