Product Details

SNP ID
rs140811887
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:919532 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTGCCACTCTGACCGCCATGAGT[C/G]TGGACCGCTGGTACGTGACGGTGTT
Phenotype
MIM: 603265 MIM: 604161
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ARID3A PubMed Links

Gene Details

Gene
ARID3A
Gene Name
AT-rich interaction domain 3A
There are no transcripts associated with this gene.

Gene
KISS1R
Gene Name
KISS1 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032551.4 1222 Missense Mutation CTG,GTG L138V NP_115940.2
XM_017027382.1 1222 Missense Mutation CTG,GTG L138V XP_016882871.1
Gene
R3HDM4
Gene Name
R3H domain containing 4
There are no transcripts associated with this gene.

View Full Product Details