Product Details

SNP ID
rs141580217
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:14799277 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTGTTCCTCAGGCTGTAGATGAAG[A/G]GGTTCAGCATGGGGGTGACCATGGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR7C1 PubMed Links

Gene Details

Gene
OR7C1
Gene Name
olfactory receptor family 7 subfamily C member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198944.1 1364 Intron NP_945182.1
XM_005259853.3 1364 Missense Mutation CCC,CTC P287L XP_005259910.1
XM_011527915.2 1364 Missense Mutation CCC,CTC P337L XP_011526217.1

View Full Product Details