Product Details

SNP ID
rs141960531
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:39386319 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGTCACTGGCCTCGTCCCTGTCA[C/T]CTTCCTCCCGTTCACTCTCGCTGCC
Phenotype
MIM: 612914 MIM: 610506
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MED29 PubMed Links

Gene Details

Gene
MED29
Gene Name
mediator complex subunit 29
There are no transcripts associated with this gene.

Gene
PAF1
Gene Name
PAF1 homolog, Paf1/RNA polymerase II complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256826.1 1599 Intron NP_001243755.1
NM_019088.3 1599 Missense Mutation GAT,GGT D423G NP_061961.2
Gene
SAMD4B
Gene Name
sterile alpha motif domain containing 4B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303614.1 1599 Intron NP_001290543.1
NM_018028.3 1599 Intron NP_060498.2
XM_011527057.2 1599 Intron XP_011525359.1
XM_011527060.2 1599 Intron XP_011525362.1
XM_011527061.2 1599 Intron XP_011525363.1
XM_011527062.2 1599 Intron XP_011525364.1
XM_011527063.2 1599 Intron XP_011525365.1
XM_011527064.2 1599 Intron XP_011525366.1
XM_017026918.1 1599 Intron XP_016882407.1
XM_017026919.1 1599 Intron XP_016882408.1
XM_017026920.1 1599 Intron XP_016882409.1
XM_017026921.1 1599 Intron XP_016882410.1
XM_017026922.1 1599 Intron XP_016882411.1
XM_017026923.1 1599 Intron XP_016882412.1

View Full Product Details