Product Details

SNP ID
rs142070691
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35778327 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGGCCCAGCCTCTCTGGAGAGAA[C/T]GAGCTGGTGTTCGGGGTGCAGGTGA
Phenotype
MIM: 614902
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARHGAP33 PubMed Links

Gene Details

Gene
ARHGAP33
Gene Name
Rho GTPase activating protein 33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172630.1 1777 UTR 5 NP_001166101.1
NM_052948.3 1777 Silent Mutation AAC,AAT N79N NP_443180.2
XM_005258488.1 1777 Silent Mutation AAC,AAT N79N XP_005258545.1
XM_006722999.1 1777 Silent Mutation AAC,AAT N79N XP_006723062.1
XM_006723003.2 1777 Intron XP_006723066.1
XM_011526416.1 1777 UTR 5 XP_011524718.1
XM_011526417.1 1777 UTR 5 XP_011524719.1
XM_011526418.1 1777 UTR 5 XP_011524720.1
XM_011526419.1 1777 UTR 5 XP_011524721.1
XM_011526420.1 1777 UTR 5 XP_011524722.1
XM_011526421.2 1777 Silent Mutation AAC,AAT N116N XP_011524723.1
XM_017026238.1 1777 Silent Mutation AAC,AAT N116N XP_016881727.1
Gene
PROSER3
Gene Name
proline and serine rich 3
There are no transcripts associated with this gene.

View Full Product Details