Product Details

SNP ID
rs142074601
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:33800520 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACACACGGCAGCACCGGCACCGC[A/G]GTGAGCCTGCAGGGTGGGCTGGGTC
Phenotype
MIM: 615240
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCTD15 PubMed Links

Gene Details

Gene
KCTD15
Gene Name
potassium channel tetramerization domain containing 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001129994.1 368 Silent Mutation GCA,GCG A22A NP_001123466.1
NM_001129995.1 368 Silent Mutation GCA,GCG A22A NP_001123467.1
NM_024076.2 368 Silent Mutation GCA,GCG A22A NP_076981.2
XM_011527296.1 368 UTR 5 XP_011525598.1
XM_011527297.2 368 Silent Mutation GCA,GCG A22A XP_011525599.1
XM_011527298.2 368 Silent Mutation GCA,GCG A22A XP_011525600.1
XM_017027283.1 368 Silent Mutation GCA,GCG A22A XP_016882772.1
XM_017027284.1 368 Intron XP_016882773.1

View Full Product Details