Product Details

SNP ID
rs142254775
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:868157 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGATCTGGGGGTCCTGGGAGAGT[A/G]GTGTGTGGACTGCGGGCCCAGCTGG
Phenotype
MIM: 134350 MIM: 604062
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CFD PubMed Links

Gene Details

Gene
CFD
Gene Name
complement factor D
There are no transcripts associated with this gene.

Gene
MED16
Gene Name
mediator complex subunit 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005481.2 2729 Missense Mutation CAC,TAC H860Y NP_005472.2
XM_017026120.1 2729 Missense Mutation CAC,TAC H791Y XP_016881609.1
XM_017026121.1 2729 Missense Mutation CAC,TAC H685Y XP_016881610.1

View Full Product Details