Product Details

SNP ID
rs142713844
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:43652309 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTCTTCAGAGGAGCATCCATGGG[C/T]GCTGTTCCCCTTGCAGCTGTAACAC
Phenotype
MIM: 173391
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLAUR PubMed Links

Gene Details

Gene
PLAUR
Gene Name
plasminogen activator, urokinase receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005376.2 718 Missense Mutation ACC,GCC T224A NP_001005376.1
NM_001005377.2 718 Missense Mutation ACC,GCC T179A NP_001005377.1
NM_001301037.1 718 Intron NP_001287966.1
NM_002659.3 718 Missense Mutation ACC,GCC T224A NP_002650.1
XM_005258990.4 718 Missense Mutation ACC,GCC T224A XP_005259047.1
XM_011527027.2 718 Missense Mutation ACC,GCC T176A XP_011525329.1
XM_011527028.2 718 Silent Mutation GCA,GCG A230A XP_011525330.1
XM_011527029.1 718 Silent Mutation GCA,GCG A230A XP_011525331.1
XM_011527030.2 718 Intron XP_011525332.1
XM_011527031.2 718 Intron XP_011525333.1
XM_017026872.1 718 Missense Mutation ACC,GCC T219A XP_016882361.1
XM_017026873.1 718 Missense Mutation ACC,GCC T100A XP_016882362.1

View Full Product Details