Product Details

SNP ID
rs142764691
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:6896333 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTCACCTCCACCCTTGTATGCTG[A/G]GTTCCCTTGCAGCCTCACTCTAATG
Phenotype
MIM: 600493
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADGRE1 PubMed Links

Gene Details

Gene
ADGRE1
Gene Name
adhesion G protein-coupled receptor E1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256252.1 Intron NP_001243181.1
NM_001256253.1 Intron NP_001243182.1
NM_001256254.1 Intron NP_001243183.1
NM_001256255.1 Intron NP_001243184.1
NM_001974.4 Intron NP_001965.3
XM_011527794.1 Intron XP_011526096.1

View Full Product Details