Product Details

SNP ID
rs142773187
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48384354 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAGATCCAGTTGAAGAGATAGAG[C/T]GTGCGGTAAACGCCTAGCGCAAACA
Phenotype
MIM: 131235 MIM: 608373
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KDELR1 PubMed Links

Gene Details

Gene
KDELR1
Gene Name
KDEL endoplasmic reticulum protein retention receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006801.2 675 Silent Mutation ACA,ACG T160T NP_006792.1
Gene
SYNGR4
Gene Name
synaptogyrin 4
There are no transcripts associated with this gene.

View Full Product Details