Product Details

SNP ID
rs142844983
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:21397249 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCCATAGCTAAGATGCCAGGACC[C/G]CCTGAAAGCCTAGACATGGTGAGAG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF493 PubMed Links

Gene Details

Gene
ZNF493
Gene Name
zinc finger protein 493
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001076678.2 131 Intron NP_001070146.1
NM_145326.2 131 Silent Mutation CCC,CCG P4P NP_663299.2
NM_175910.6 131 UTR 5 NP_787106.4
Gene
ZNF738
Gene Name
zinc finger protein 738
There are no transcripts associated with this gene.

View Full Product Details