Product Details

SNP ID
rs142961225
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:8316635 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAGGACCCACAGGGAGCTTGGGAG[A/G]AGGCTGAGTTCTGAGGGGAAAAAAG
Phenotype
MIM: 606475 MIM: 614611 MIM: 602139 MIM: 603685
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD320 PubMed Links

Gene Details

Gene
CD320
Gene Name
CD320 molecule
There are no transcripts associated with this gene.

Gene
KANK3
Gene Name
KN motif and ankyrin repeat domains 3
There are no transcripts associated with this gene.

Gene
NDUFA7
Gene Name
NADH:ubiquinone oxidoreductase subunit A7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005001.4 167 Missense Mutation CCT,TCT P38S NP_004992.2
Gene
RPS28
Gene Name
ribosomal protein S28
There are no transcripts associated with this gene.

View Full Product Details