Product Details

SNP ID
rs143150861
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:19121545 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAACTGCAGCATGGGTCTGTCCTC[C/T]GACATGGTCAGTGCGTCCCGGTGGG
Phenotype
MIM: 610823
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC25A42 PubMed Links

Gene Details

Gene
SLC25A42
Gene Name
solute carrier family 25 member 42
There are no transcripts associated with this gene.

Gene
TMEM161A
Gene Name
transmembrane protein 161A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256766.1 749 Silent Mutation TCA,TCG S157S NP_001243695.1
NM_017814.2 749 Silent Mutation TCA,TCG S260S NP_060284.1
XM_011528117.2 749 Silent Mutation TCA,TCG S58S XP_011526419.1
XM_017026916.1 749 Silent Mutation TCA,TCG S235S XP_016882405.1

View Full Product Details