Product Details

SNP ID
rs143353680
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11169880 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCGGCCGGATTGAGACTCACCGA[A/G]CACTTGATGTTCATGCGGGAATACA
Phenotype
MIM: 614610
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KANK2 PubMed Links

Gene Details

Gene
KANK2
Gene Name
KN motif and ankyrin repeat domains 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136191.2 2860 Silent Mutation TGC,TGT C833C NP_001129663.1
NM_015493.6 2860 Silent Mutation TGC,TGT C841C NP_056308.3

View Full Product Details