Product Details

SNP ID
rs143781796
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:19629653 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTGTCCCAGGTGTCAGAGACTAG[A/G]GTGGACCCCAAGGATCTCATAACCT
Phenotype
MIM: 609694 MIM: 605110
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GMIP PubMed Links

Gene Details

Gene
GMIP
Gene Name
GEM interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288998.1 3274 UTR 3 NP_001275927.1
NM_001288999.1 3274 UTR 3 NP_001275928.1
NM_016573.3 3274 UTR 3 NP_057657.2
XM_005259927.2 3274 UTR 3 XP_005259984.1
XM_011528048.2 3274 UTR 3 XP_011526350.1
XM_017026861.1 3274 UTR 3 XP_016882350.1
XM_017026862.1 3274 Intron XP_016882351.1
Gene
LPAR2
Gene Name
lysophosphatidic acid receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004720.5 3274 Intron NP_004711.2
XM_011528421.2 3274 Intron XP_011526723.1
XM_017027468.1 3274 Intron XP_016882957.1
XM_017027469.1 3274 Intron XP_016882958.1

View Full Product Details