Product Details

SNP ID
rs143979135
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48048780 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATCCAGCAATAACAGCTTTCCTTC[C/T]GTTCCTCGAATAGACTGATATAGTT
Phenotype
MIM: 607315 MIM: 603602
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CABP5 PubMed Links

Gene Details

Gene
CABP5
Gene Name
calcium binding protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019855.4 Intron NP_062829.1
XM_017026992.1 Intron XP_016882481.1
Gene
PLA2G4C
Gene Name
phospholipase A2 group IVC
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159322.1 Intron NP_001152794.1
NM_001159323.1 Intron NP_001152795.1
NM_003706.2 Intron NP_003697.2
XM_011527431.2 Intron XP_011525733.1
XM_011527432.2 Intron XP_011525734.1
XM_011527433.2 Intron XP_011525735.1
XM_017027411.1 Intron XP_016882900.1
XM_017027412.1 Intron XP_016882901.1
XM_017027413.1 Intron XP_016882902.1
XM_017027414.1 Intron XP_016882903.1

View Full Product Details