Product Details

SNP ID
rs144141559
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:43795834 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTGCTATGCACTTACTCTGAAAA[C/T]TGCTCTCTCACTGTGCTTTGGGCAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LYPD5 PubMed Links

Gene Details

Gene
LYPD5
Gene Name
LY6/PLAUR domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031749.2 2650 Intron NP_001026919.2
NM_001288763.1 2650 Intron NP_001275692.1
NM_182573.2 2650 Intron NP_872379.2
XM_011526794.2 2650 UTR 3 XP_011525096.1

View Full Product Details