Product Details

SNP ID
rs144586938
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57573129 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCCAGGGTGGACTCTTTGCAGCT[A/G]AACAAGGGAGCACTCACAGCAGCAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF416 PubMed Links

Gene Details

Gene
ZNF416
Gene Name
zinc finger protein 416
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017879.1 946 Nonsense Mutation CAG,TAG Q259* NP_060349.1
XM_005259051.3 946 Nonsense Mutation CAG,TAG Q187* XP_005259108.1

View Full Product Details