Product Details

SNP ID
rs144613276
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10041510 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCTCACCCCGGCCCCTGACTGAG[A/G]TCAGGACATGGTGTTCTTGCCCAAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C3P1 PubMed Links

Gene Details

Gene
C3P1
Gene Name
complement component 3 precursor pseudogene
There are no transcripts associated with this gene.

Gene
MIR5589
Gene Name
microRNA 5589
There are no transcripts associated with this gene.

View Full Product Details