Product Details

SNP ID
rs144874497
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57154468 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACTAGTGAAGTTGGTGCCATTTT[C/G]TGTATCTTCTGCACCTAAGGAGGAA
Phenotype
MIM: 611168
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DUXA PubMed Links

Gene Details

Gene
DUXA
Gene Name
double homeobox A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012729.1 604 Missense Mutation CAA,GAA Q187E NP_001012747.1
Gene
ZIM3
Gene Name
zinc finger imprinted 3
There are no transcripts associated with this gene.

View Full Product Details