Product Details

SNP ID
rs145238835
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17872649 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTGCTCTTCATGCCCGTCTTCTA[C/T]CGCCTGGGCCTCACCAGCACCTACG
Phenotype
MIM: 604178 MIM: 601843
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RPL18A PubMed Links

Gene Details

Gene
RPL18A
Gene Name
ribosomal protein L18a
There are no transcripts associated with this gene.

Gene
SLC5A5
Gene Name
solute carrier family 5 member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000453.2 1256 Silent Mutation TAC,TAT Y110Y NP_000444.1
XM_011528192.2 1256 Silent Mutation TAC,TAT Y110Y XP_011526494.1
XM_011528193.2 1256 Silent Mutation TAC,TAT Y21Y XP_011526495.1
XM_011528194.2 1256 Intron XP_011526496.1
XM_017027158.1 1256 Silent Mutation TAC,TAT Y21Y XP_016882647.1
Gene
SNORA68
Gene Name
small nucleolar RNA, H/ACA box 68
There are no transcripts associated with this gene.

View Full Product Details