Product Details

SNP ID
rs145372180
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54462294 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCTGGAGACTGTAGTGTACTCAG[C/T]CCCTCGGCTTCCAGCCTCTGGCTCA
Phenotype
MIM: 609171 MIM: 616575
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDC42EP5 PubMed Links

Gene Details

Gene
CDC42EP5
Gene Name
CDC42 effector protein 5
There are no transcripts associated with this gene.

Gene
LENG8
Gene Name
leukocyte receptor cluster member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052925.3 1421 Intron NP_443157.1
XM_005278248.4 1421 Intron XP_005278305.1
XM_005278249.4 1421 Intron XP_005278306.1
XM_005278250.4 1421 Intron XP_005278307.1
XM_005278252.4 1421 Intron XP_005278309.1
XM_006722996.3 1421 Intron XP_006723059.1
XM_006722997.3 1421 Intron XP_006723060.1
XM_011526413.2 1421 Intron XP_011524715.1
XM_011526414.2 1421 Intron XP_011524716.1
XM_011526415.2 1421 Intron XP_011524717.1
Gene
LENG9
Gene Name
leukocyte receptor cluster member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301782.1 1421 Silent Mutation GGA,GGG G411G NP_001288711.1
NM_198988.1 1421 Silent Mutation GGA,GGG G434G NP_945339.2

View Full Product Details