Product Details

SNP ID
rs145395343
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:9093738 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCACTCACATCCTCCTGATGGCCC[A/G]TCTCGTTTTCTGCGGCAGCCATGAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR1M1 PubMed Links

Gene Details

Gene
OR1M1
Gene Name
olfactory receptor family 1 subfamily M member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004456.1 494 Missense Mutation CAT,CGT H165R NP_001004456.1
Gene
OR7G2
Gene Name
olfactory receptor family 7 subfamily G member 2
There are no transcripts associated with this gene.

View Full Product Details