Product Details

SNP ID
rs146125110
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:58388224 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGTGGAGCACCGATGATGTGCA[A/G]ATCAATGACATTTCCCTGCAGGTGA
Phenotype
MIM: 603630
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR4754 PubMed Links

Gene Details

Gene
MIR4754
Gene Name
microRNA 4754
There are no transcripts associated with this gene.

Gene
RNF225
Gene Name
ring finger protein 225
There are no transcripts associated with this gene.

Gene
RPS5
Gene Name
ribosomal protein S5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009.3 Intron NP_001000.2
Gene
ZNF837
Gene Name
zinc finger protein 837
There are no transcripts associated with this gene.

View Full Product Details