Product Details

SNP ID
rs146415600
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:58498570 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGTACATTTCTGCCGTCAGGGGC[C/T]GGAAGGACTGGGCCCGGTTGTCCAG
Phenotype
MIM: 603314
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC27A5 PubMed Links

Gene Details

Gene
SLC27A5
Gene Name
solute carrier family 27 member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321196.1 2128 Missense Mutation CAG,CGG Q589R NP_001308125.1
NM_012254.2 2128 Missense Mutation CAG,CGG Q673R NP_036386.1
XM_011526364.1 2128 UTR 3 XP_011524666.1
XM_017026214.1 2128 Intron XP_016881703.1
Gene
ZNF446
Gene Name
zinc finger protein 446
There are no transcripts associated with this gene.

View Full Product Details