Product Details

SNP ID
rs146428235
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:40472692 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCCTAACAACAACCCTGCTGCCC[G/T]CTGGGAGAGTCCGGATCGGGGCTGG
Phenotype
MIM: 600941 MIM: 606214
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
BLVRB PubMed Links

Gene Details

Gene
BLVRB
Gene Name
biliverdin reductase B
There are no transcripts associated with this gene.

Gene
SPTBN4
Gene Name
spectrin beta, non-erythrocytic 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020971.2 239 Missense Mutation CGC,CTC R24L NP_066022.2
NM_025213.2 239 Intron NP_079489.2
XM_011527173.2 239 Missense Mutation CGC,CTC R24L XP_011525475.1
XM_017027049.1 239 Missense Mutation CGC,CTC R24L XP_016882538.1
XM_017027050.1 239 Intron XP_016882539.1
XM_017027051.1 239 Missense Mutation CGC,CTC R24L XP_016882540.1
XM_017027052.1 239 Missense Mutation CGC,CTC R24L XP_016882541.1

View Full Product Details