Product Details

SNP ID
rs146522321
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:547378 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGAGAAATGGCTCCCACCTGTGC[C/G]GGGGTGTCCTGGTGCACCCAAAGTG
Phenotype
MIM: 116948 MIM: 600311
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CDC34 PubMed Links

Gene Details

Gene
CDC34
Gene Name
cell division cycle 34
There are no transcripts associated with this gene.

Gene
GZMM
Gene Name
granzyme M
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258351.1 192 Missense Mutation CGG,GGG R13G NP_001245280.1
NM_005317.3 192 Missense Mutation CGG,GGG R52G NP_005308.1

View Full Product Details