Product Details

SNP ID
rs146845211
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:39825922 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACCAGGCTCACATCCATCAGCTC[C/T]GGGGGTGGTGGTGAGGTTGGTGATG
Phenotype
MIM: 604556 MIM: 134795
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DYRK1B PubMed Links

Gene Details

Gene
DYRK1B
Gene Name
dual specificity tyrosine phosphorylation regulated kinase 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004714.2 1911 Silent Mutation CCA,CCG P561P NP_004705.1
NM_006483.2 1911 Silent Mutation CCA,CCG P521P NP_006474.1
NM_006484.2 1911 Silent Mutation CCA,CCG P533P NP_006475.1
XM_005259398.4 1911 Silent Mutation CCA,CCG P561P XP_005259455.1
Gene
FBL
Gene Name
fibrillarin
There are no transcripts associated with this gene.

Gene
MIR6719
Gene Name
microRNA 6719
There are no transcripts associated with this gene.

View Full Product Details