Product Details

SNP ID
rs147003075
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12349786 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCTTCCCACATTCCTTACATTCA[C/T]GCATCTTCTCTCCAGTGTGAGTTTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF442 PubMed Links

Gene Details

Gene
ZNF442
Gene Name
zinc finger protein 442
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030824.2 1838 Missense Mutation CAT,CGT H600R NP_110451.1
XM_006722908.3 1838 Missense Mutation CAT,CGT H593R XP_006722971.1
XM_017027316.1 1838 Missense Mutation CAT,CGT H608R XP_016882805.1
XM_017027317.1 1838 Missense Mutation CAT,CGT H543R XP_016882806.1
XM_017027318.1 1838 Missense Mutation CAT,CGT H600R XP_016882807.1
Gene
ZNF563
Gene Name
zinc finger protein 563
There are no transcripts associated with this gene.

View Full Product Details