Product Details
- SNP ID
-
rs147202003
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:57926261 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTCATAAGGCCTTTCTCCAGTGTG[A/T]ACTCTCCTGTGTTCAGTAAGACTGA
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF417
PubMed Links
Gene Details
- Gene
- ZNF417
- Gene Name
- zinc finger protein 417
There are no transcripts associated with this gene.
- Gene
- ZNF418
- Gene Name
- zinc finger protein 418
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001317027.1 |
2226 |
Silent Mutation |
GTA,GTT |
V661V |
NP_001303956.1 |
NM_001317028.1 |
2226 |
Silent Mutation |
GTA,GTT |
V641V |
NP_001303957.1 |
NM_001317029.1 |
2226 |
Silent Mutation |
GTA,GTT |
V640V |
NP_001303958.1 |
NM_001317030.1 |
2226 |
Silent Mutation |
GTA,GTT |
V555V |
NP_001303959.1 |
NM_133460.2 |
2226 |
Silent Mutation |
GTA,GTT |
V640V |
NP_597717.1 |
XM_017026304.1 |
2226 |
Silent Mutation |
GTA,GTT |
V661V |
XP_016881793.1 |
XM_017026305.1 |
2226 |
Silent Mutation |
GTA,GTT |
V661V |
XP_016881794.1 |
XM_017026306.1 |
2226 |
Silent Mutation |
GTA,GTT |
V660V |
XP_016881795.1 |
XM_017026307.1 |
2226 |
Silent Mutation |
GTA,GTT |
V650V |
XP_016881796.1 |
XM_017026308.1 |
2226 |
Silent Mutation |
GTA,GTT |
V650V |
XP_016881797.1 |
XM_017026309.1 |
2226 |
Silent Mutation |
GTA,GTT |
V641V |
XP_016881798.1 |
XM_017026310.1 |
2226 |
Silent Mutation |
GTA,GTT |
V641V |
XP_016881799.1 |
XM_017026311.1 |
2226 |
Silent Mutation |
GTA,GTT |
V641V |
XP_016881800.1 |
XM_017026312.1 |
2226 |
Silent Mutation |
GTA,GTT |
V640V |
XP_016881801.1 |
XM_017026313.1 |
2226 |
Silent Mutation |
GTA,GTT |
V640V |
XP_016881802.1 |
XM_017026314.1 |
2226 |
Silent Mutation |
GTA,GTT |
V607V |
XP_016881803.1 |
View Full Product Details